Calcium oxalate monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxalic acid calcium salt monohydrate, Calcium ethanedioate monohydrate

032430-22 032430-36
AA32430-22EA 57.1 USD
AA32430-22 AA32430-36
Calcium oxalate monohydrate 99%
Calcium oxalate monohydrate

MDL: MFCD00150039


Purity: 99% CAS No.: 5794-28-5 Molecular Formula: CaC2O4.H2O Formula Weight: 146.11 (128.10anhy) Physical Form: Powder/lump Density: 2.12 Melting Point: 200° Merck Reference: 14,1685 UN No.: 3288 MDL No.: MFCD00150039

Formula: C₂CaO₄·1H₂O
MW: 146.11 g/mol
Melting Pt: 200 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150039
CAS Number: 5794-28-5
EINECS: 209-260-1
UN: 3288
ADR: 6.1,III
Merck Index: 14,1685

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR