Gallium trinitrate hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

032116-06 032116-14
AA32116-06EA 66.51 USD
AA32116-06 AA32116-14
Gallium trinitrate hydrate ≥99.9% (metals basis)
Gallium trinitrate hydrate

MDL: MFCD00149723


Purity: 99.9% CAS No.: 69365-72-6 Molecular Formula: GaN3O9.xH2O Formula Weight: 255.73(anhy) Physical Form: Lump Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 110° dec. Merck Reference: 14,4346 UN No.: 1477 MDL No.: MFCD00149723

Formula: GaN₃O₉·H₂O
MW: 273.75
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1 III
Merck Index: 14,4346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR