n-Hexatriacontane 97+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Hexatriacontane
031641-09
031641-18
AA31641-09EA
58.5
USD
AA31641-09
AA31641-18
n-Hexatriacontane 97+%
n-Hexatriacontane
MDL: MFCD00009413
Purity: 97+%
CAS No.: 630-06-8
Molecular Formula: CH3(CH2)34CH3
Formula Weight: 506.95
Physical Form: Wax
Density: 0.779
Refractive Index: 1.4573
Appearance: Off-white
Melting Point: 76.2°
Boiling Point: 497°
MDL No.: MFCD00009413
Formula:
H₃C(CH₂)₃₄CH₃ MW: 506.98 g/mol Boiling Pt: 497 °C Melting Pt: 76.2 °C Density: 0.7803 g/cm³ (80 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009413 CAS Number: 630-06-8 EINECS: 211-127-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...