Dimethylamine 40% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

031458-A1 031458-30 031458-A5
AA31458-A1EA 126 USD
AA31458-A1 AA31458-30 AA31458-A5
Dimethylamine 40% (w/w) in aqueous solution
Dimethylamine

MDL: MFCD00008288 Beilstein Registry No.: 605257


CAS No.: 124-40-3 Molecular Formula: (CH3)2NH Formula Weight: 45.08 Flash Point: 15°(60°F) Physical Form: Liquid Density: 0.890 Refractive Index: 1.3700 Odor: Fish-like Beilstein Reference: 605257 Merck Reference: 14,3228 UN No.: 1160 MDL No.: MFCD00008288

Formula: C₂H₇N
MW: 45.08 g/mol
Density: 0.890
Flash Pt: 15 °C (60 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3,II
Merck Index: 14,3228

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR