Tetramethylammonium perchlorate
Supplier: Thermo Scientific Chemicals
|
Danger
|
030834-22
030834-14
AA30834-22EA
363
USD
AA30834-22
AA30834-14
Tetramethylammonium perchlorate
Tetramethylammonium perchlorate
MDL: MFCD00043207
CAS No.: 2537-36-2
Molecular Formula: (CH3)4NClO4
Formula Weight: 173.60
Physical Form: Crystalline
Melting Point:
Formula:
C₄H₁₂ClNO₄ Melting Pt: >300 °C Density: 1.379 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00043207 CAS Number: 2537-36-2 EINECS: 219-805-5 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...