Tetrabutylammonium perchlorate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetra-n-butylammonium perchlorate

030801-14 030801-22 030801-88
AA30801-14EA 126.32 USD
AA30801-14 AA30801-22 AA30801-88
Tetrabutylammonium perchlorate 99+%
Tetrabutylammonium perchlorate

MDL: MFCD00038722


Purity: 99+% CAS No.: 1923-70-2 Molecular Formula: C16H36ClNO4 Formula Weight: 341.92 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: 210-217° UN No.: 1481 MDL No.: MFCD00038722

Formula: C₁₆H₃₆ClNO₄
MW: 341.92 g/mol
Melting Pt: 210…217 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00038722
CAS Number: 1923-70-2
EINECS: 217-655-5
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR