Thioacetamide ≥99% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethanethioamide

030638-18 030638-30 030638-09
AA30638-18EA 162.2 USD
AA30638-18 AA30638-30 AA30638-09
Thioacetamide ≥99% ACS
Thioacetamide

MDL: MFCD00008070 Beilstein Registry No.: 506006 Fieser: 4,502


Purity: 99% CAS No.: 62-55-5 Molecular Formula: CH3CSNH2 Formula Weight: 75.13 Physical Form: Crystalline Melting Point: 112-114° Beilstein Reference: 506006 Merck Reference: 14,9319 MDL No.: MFCD00008070

Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 112…114 °C
Density: 1.336 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Residue after ignition 0.05%
Clarity of a 2% solution Pass test.
Melting point 111-114°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR