Tricyclohexylphosphine ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
030386-14
030386-03
030386-06
AA30386-14EA
365.12
USD
AA30386-14
AA30386-03
AA30386-06
Tricyclohexylphosphine ≥96%
Tricyclohexylphosphine
MDL: MFCD00003853
Purity: 96%
CAS No.: 2622-14-2
Molecular Formula: (C6H11)3P
Formula Weight: 280.42
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White to off-white
Odor: Unpleasant
Melting Point: 76-78°
MDL No.: MFCD00003853
Formula:
C₁₈H₃₃P Boiling Pt: 132…134 °C (0.02 mmHg) Melting Pt: 76…78 °C Density: 0.9 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...