Cobalt(II) oxide ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cobalt oxide
023141-22
023141-14
AA23141-22EA
351
USD
AA23141-22
AA23141-14
Cobalt(II) oxide ≥95%
Cobalt(II) oxide
Pigments, glass coloring, semiconductors
Purity: 95%
CAS No.: 1307-96-6
Molecular Formula: CoO
Formula Weight: 74.93
Physical Form: Powder
Density: 6.45
Appearance: Black-green
Melting Point: 1935°
Solubility: Soluble in acids or alkalies. Insoluble in water
Merck Reference: 14,2446
UN No.: 2811
MDL No.: MFCD00016031
Pigments, glass coloring, semiconductors
Formula:
CoO MW: 74.93 g/mol Melting Pt: 1933 Density: 6.45 Storage Temperature: Ambient |
MDL Number:
MFCD00016031 CAS Number: 1307-96-6 EINECS: 215-154-6 UN: 2811 ADR: 6.1 III Merck Index: 14,2446 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...