Cobalt(II) acetate Co ≥ 32%, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobaltous acetate, Cobalt acetate, Acetic acid cobalt (II) salt, Cobalt diacetate

023138-14 023138-22
AA23138-14EA 257.17 USD
AA23138-14 AA23138-22
Cobalt(II) acetate Co ≥ 32%, anhydrous
Cobalt(II) acetate

MDL: MFCD00008689 Soluble in water


CAS No.: 71-48-7 Molecular Formula: C4H6CoO4 Formula Weight: 177.02 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Pink Odor: Odorless Melting Point: 298° dec. Solubility: Soluble in water Merck Reference: 14,2433 UN No.: 3077 MDL No.: MFCD00008689

Formula: C₄H₆CoO₄
MW: 177.02 g/mol
Melting Pt: 298 °C (dec.)
Density: 1.7755 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008689
CAS Number: 71-48-7
EINECS: 200-755-8
UN: 3077
ADR: 9 III
Merck Index: 14,2433

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR