N,N-Dimethylacetamide ≥99.5% for HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid dimethylamide, Dimethyl Acetamide, DMAC
022916-K2
022916-K7
022916-M1
AA22916-K2EA
164.8
USD
AA22916-K2
AA22916-K7
AA22916-M1
N,N-Dimethylacetamide ≥99.5% for HPLC
N,N-Dimethylacetamide
MDL: MFCD00008686
Beilstein Registry No.: 1737614
Notes: Filtered through 0.2µ filters.
Fieser: 1,270 2,144 4,165
Purity: 99.5+%
CAS No.: 127-19-5
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 0.940
Refractive Index: 1.4385
Appearance: Colorless
Odor: Ammonia-like
Melting Point: -20°
Boiling Point: 165-166°
Beilstein Reference: 1737614
Merck Reference: 14,3227
MDL No.: MFCD00008686
Formula:
CH₃CON(CH₃)₂ MW: 87.12 g/mol Boiling Pt: 166.1 Melting Pt: -20 Density: 0.940 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 UN: 2810 ADR: 6.1 III Merck Index: 14,3227 |
|
Specification Test Results
Evaporation residue | 5 ppm |
H2O | 0.0005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...