Copper(II) acetate, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid copper (II) salt, Cupric acetate, Copper diacetate

019417-22 019417-36 019417-14
AA19417-22EA 180 USD
AA19417-22 AAA19417-36 AA19417-14
Copper(II) acetate, anhydrous
Copper(II) acetate

Anhydrous


CAS No.: 142-71-2 Molecular Formula: Cu(OOCCH3)2 Formula Weight: 181.63 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Blue-green Odor: Odorless Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00008690

Formula: CuCH₃CO₂
MW: 181.64 g/mol
Boiling Pt: 240
Melting Pt: 115
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008690
CAS Number: 142-71-2
EINECS: 205-553-3
UN: 3077
ADR: 9 III
Merck Index: 14,2624

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR