2-Methylbutane 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
iso-Pentane
019387-AP
019387-AY
AA19387-APEA
74.52
USD
AA19387-AP
AA19387-AY
2-Methylbutane 99+%
2-Methylbutane
MDL: MFCD00009338
Purity: 99+%
CAS No.: 78-78-4
Molecular Formula: C5H12
Formula Weight: 72.14
Flash Point: -57°(-70°F)
Physical Form: Liquid
Density: 0.62
Refractive Index: 1.3540
Appearance: Colorless
Boiling Point: 30°
UN No.: 1265
MDL No.: MFCD00009338
Formula:
CH₃CH₂CH(CH₃)₂ MW: 72.15 g/mol Boiling Pt: 28 Melting Pt: -160 Density: 0.62 Flash Pt: –57 °C (–70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009338 CAS Number: 78-78-4 EINECS: 201-142-8 UN: 1265 ADR: 3 I |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...