Tetrapropylammonium chloride 99+%

Supplier: Thermo Scientific Chemicals
Warning

019379-06 019379-03 019379-14
AA19379-06EA 41.4 USD
AA19379-06 AA19379-03 AA19379-14
Tetrapropylammonium chloride 99+%
Tetrapropylammonium chloride

MDL: MFCD00038729 Soluble in water, acetone


Purity: 99+% CAS No.: 5810-42-4 Molecular Formula: C12H28ClN Formula Weight: 221.87 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 240-242° Solubility: Soluble in water, acetone MDL No.: MFCD00038729

Formula: C₁₂H₂₈ClN
Melting Pt: 240…242 °C
Density: 1.0334 g/cm³ (3 °C)
Storage Temperature: Ambient
MDL Number: MFCD00038729
CAS Number: 5810-42-4
EINECS: 227-375-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR