Tetracyclohexyltin
Supplier: Thermo Scientific Chemicals
|
Warning
|
018822-06
018822-14
018822-03
AA18822-06EA
342
USD
AA18822-06
AA18822-14
AA18822-03
Tetracyclohexyltin
Tetracyclohexyltin
MDL: MFCD00014278
CAS No.: 1449-55-4
Molecular Formula: C24H44Sn
Formula Weight: 451.31
Physical Form: Powder
Melting Point: 241-250°
UN No.: 3146
MDL No.: MFCD00014278
Formula:
C₂₄H₄₄Sn MW: 451.32 g/mol Melting Pt: 241…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014278 CAS Number: 1449-55-4 EINECS: 215-910-5 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...