Calcium bromide hydrate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

018820-06 018820-22 018820-36
AA18820-06EA 42 USD
AA18820-06 AA18820-22 AA18820-36
Calcium bromide hydrate ≥95%
Calcium bromide hydrate

MDL: MFCD00149608


Purity: 95% CAS No.: 71626-99-8 Molecular Formula: CaBr2.xH2O Formula Weight: 199.90(anhy) Physical Form: Powder Appearance: White MDL No.: MFCD00149608

Formula: Br₂Ca·H₂O
Density: 2,295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149608
CAS Number: 71626-99-8
EINECS: 232-164-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR