1,5-Hexadiyne 50% in pentane
Supplier: Thermo Scientific Chemicals
|
Danger
|
018720-09
018720-04
AA18720-09EA
444
USD
AA18720-09
AA18720-04
1,5-Hexadiyne 50% in pentane
1,5-Hexadiyne
MDL: MFCD00014924
CAS No.: 628-16-0
Molecular Formula: HC≡CCH2CH2C≡CH
Formula Weight: 78.11
Flash Point: -2°(29°F)
Physical Form: Liquid, Packaged under argon
Density: 0.805
UN No.: 3295
MDL No.: MFCD00014924
Formula:
C₆H₆ Density: 0.805 Flash Pt: –2 °C (29 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014924 CAS Number: 628-16-0 EINECS: 211-029-5 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...