Neodymium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Neodymium (III) chloride

018680-06 018680-22 018680-14
AA18680-06EA 90.62 USD
AA18680-06 AA18680-22 AA18680-14
Neodymium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Neodymium trichloride

MDL: MFCD00011132


Purity: 99.9% CAS No.: 10024-93-8 Molecular Formula: NdCl3 Formula Weight: 250.60 Physical Form: -20 Mesh Powder Density: 4.134 Melting Point: 784° Merck Reference: 14,6450 MDL No.: MFCD00011132

Formula: Cl₃Nd
MW: 250.6 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 784 °C
Density: 4.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5
Merck Index: 14,6450

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR