Scandium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
Warning
|
018677-03
018677-06
018677-01
AA18677-03EA
243
USD
AA18677-03
AA18677-06
AA18677-01
Scandium(III) chloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Scandium(III) chloride
MDL: MFCD00011221
Notes: Contains 5-10% oxy compounds
Soluble in water. Insoluble in alcohol
Purity: 99.9%
CAS No.: 10361-84-9
Molecular Formula: ScCl3
Formula Weight: 151.32
Physical Form: -20 Mesh
Density: 2.39
Melting Point: 939°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,8392
MDL No.: MFCD00011221
Formula:
Cl₃Sc MW: 151.31 g/mol Melting Pt: 939 °C Density: 1.67 g/cm³ (940 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011221 CAS Number: 10361-84-9 EINECS: 233-799-1 Merck Index: 14,8392 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...