Indium tin alloy (52:48 w%), eutectic ≥99.99% (metals basis), ingot
Supplier: Thermo Scientific Chemicals
018159-50
018159-09
AA18159-50EA
336
USD
AA18159-50
AA18159-09
Indium tin alloy (52:48 w%), eutectic ≥99.99% (metals basis), ingot
Indium tin alloy (52:48 w%), eutectic
MDL: MFCD00144477
Purity: 99.99%
Molecular Formula: In:Sn; 52:48 wt%
Melting Point: 117°
MDL No.: MFCD00144477
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...