Styrene ≥99.5% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylethene, Phenylethylene, Vinylbenzene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
MDL: MFCD00008612
Beilstein Registry No.: 1071236
Fieser: 1,935 4,409 5,557 6,486 7,303 8,415 9,387 11,439 21,360
Purity: 99.5%
CAS No.: 100-42-5
Molecular Formula: C8H8
Formula Weight: 104.15
Storage Temperature: 2°C to 8°C
Flash Point: 31°(88°F)
Physical Form: Liquid
Density: 0.906
Refractive Index: 1.5460
Sensitivity: Light Sensitive
Appearance: Colorless
Odor: Pungent
pH: neutral
Melting Point: -31°
Boiling Point: 145-146°
Beilstein Reference: 1071236
Merck Reference: 14,8860
UN No.: 2055
MDL No.: MFCD00008612
Formula:
C₈H₈ MW: 104.15 g/mol Boiling Pt: 145…146 °C Melting Pt: –31 °C Density: 0.906 g/cm³ (20 °C) Flash Pt: 31 °C (88 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008612 CAS Number: 100-42-5 EINECS: 202-851-5 UN: 2055 ADR: 3,III Merck Index: 13,08944 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...