Indium ≥99.999% (metals basis), ingot

Supplier: Thermo Scientific Chemicals

014720-18 014720-09
AA14720-18EA 354.07 USD
AA14720-18 AA14720-09
Indium ≥99.999% (metals basis), ingot
Indium

MDL: MFCD00134048


Purity: 99.999% CAS No.: 7440-74-6 Physical Form: Soft white ingot Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR