Ammonium iodate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium Iodate, Iodic acid, ammonium salt

014531-18 014531-30
AA14531-18EA 65.4 USD
AA14531-18 AA14531-30
Ammonium iodate 99%
Ammonium iodate

MDL: MFCD00049684


Purity: 99% CAS No.: 13446-09-8 Molecular Formula: H4INO3 Formula Weight: 192.94 Physical Form: Crystalline Density: 3.309 Appearance: White Odor: Odorless Melting Point: 150° UN No.: 1479 MDL No.: MFCD00049684

Formula: H₄INO₃
MW: 192.94 g/mol
Melting Pt: 150 °C
Density: 3.407 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049684
CAS Number: 13446-09-8
EINECS: 236-592-4
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR