Mercury(II) nitrate monohydrate ≥98.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury(II) nitrate hydrate, Mercury dinitrate monohydrate

014497-18 014497-30
AA14497-18EA 93.87 USD
AA14497-18 AA14497-30
Mercury(II) nitrate monohydrate ≥98.0% ACS
Mercury(II) nitrate monohydrate

MDL: MFCD00149736


Purity: 98.0% min CAS No.: 7783-34-8 Molecular Formula: HgN2O6.xH2O (x=1-2) Formula Weight: 324.60(anhy) Physical Form: Crystalline Density: 4.39 Appearance: White Melting Point: 79° Merck Reference: 14,5880 UN No.: 1625 MDL No.: MFCD00149736

Formula: Hg(NO₃)₂·1H₂O
MW: 342.62 g/mol
Melting Pt: 79
Density: 4.39
Storage Temperature: Ambient
MDL Number: MFCD00149736
CAS Number: 7783-34-8
EINECS: 233-152-3
UN: 1625
ADR: 6.1 II
Merck Index: 14,5880

Order Now

Specification Test Results

Residue after reduction 0.01%
Cl 0.002%
SO3 0.002%
Fe 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR