Calcium ≥99.98% (metals basis), crystalline dendritic pieces

Supplier: Thermo Scientific Chemicals
Danger

014457-06 014457-14
AA14457-06EA 285 USD
AA14457-06 AA14457-14
Calcium ≥99.98% (metals basis), crystalline dendritic pieces
Calcium

MDL: MFCD00085314


Purity: 99.98% CAS No.: 7440-70-2 Physical Form: Ampouled under argon Odor: Odorless UN No.: 1401 MDL No.: MFCD00085314

Formula: Ca
Boiling Pt: 1484 °C (1013 hPa)
Melting Pt: 850 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR