Manganese(II) carbonate ≥99.9% (metals basis excluding Na)
Supplier: Thermo Scientific Chemicals
Synonyms:
Manganous carbonate
014344-30
014344-A1
AA14344-30EA
72.03
USD
AA14344-30
AA14344-A1
Manganese(II) carbonate ≥99.9% (metals basis excluding Na)
Manganese(II) carbonate
MDL: MFCD00011116
Purity: 99.9%
CAS No.: 598-62-9
Molecular Formula: MnCO3
Formula Weight: 114.95
Physical Form: Powder
Density: 3.125
Appearance: Light brown to violet
Odor: Odorless
Melting Point: 350° dec.
Merck Reference: 14,5726
MDL No.: MFCD00011116
Formula:
CMnO₃ MW: 114.95 g/mol Melting Pt: 350 °C (dec.) Density: 3.125 Storage Temperature: Ambient |
MDL Number:
MFCD00011116 CAS Number: 598-62-9 EINECS: 209-942-9 Merck Index: 14,5726 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...