Potassium nitrite ≥96.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

014245-09 014245-22 014245-36
AA14245-09EA 32.6 USD
AA14245-09 AA14245-22 AA14245-36
Potassium nitrite ≥96.0% ACS
Potassium nitrite

In analytical chemistry (testing for amino acids, cobalt, iodine, urea)


Purity: 96.0% min CAS No.: 7758-09-0 Molecular Formula: KNO2 Formula Weight: 85.11 Physical Form: Solid Density: 1.915 Appearance: White to pale yellow Odor: Odorless pH: 7.0-10.0 Melting Point: 350° dec. Solubility: Soluble in water, hot alcohol, liquid NH3 Merck Reference: 14,7649 UN No.: 1488 MDL No.: MFCD00011408


In analytical chemistry (testing for amino acids, cobalt, iodine, urea)

Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 350 °C (dec.)
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 13,07734

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.03%
SO3 Pass test
Ca 0.005%
MG 0.002%
Heavy Metals (as Pb) 0.001%
Fe 0.001%
NA 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR