Sodium nitrite ≥97% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011118
Notes: Decomposes above 320°C
Soluble in water
Fieser: 1,1097 11,491 13,282 16,308 18,336 19,313 20,350 21,405
Purity: 97% min
CAS No.: 7632-00-0
Molecular Formula: NaNO2
Formula Weight: 69.00
Physical Form: Granular
Density: 2.17
Melting Point: 271°
Solubility: Soluble in water
Merck Reference: 14,8648
UN No.: 1500
MDL No.: MFCD00011118
Formula:
NaNO₂ Boiling Pt: 320 °C (1013 hPa) Melting Pt: 271 °C Density: 2.17 Storage Temperature: Ambient |
MDL Number:
MFCD00011118 CAS Number: 7632-00-0 EINECS: 231-555-9 UN: 1500 ADR: 5.1,III Merck Index: 14,8648 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.005% |
SO3 | 0.01% |
Ca | 0.01% |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.001% |
K | 0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...