Lead(II,IV) oxide ≥97% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Minium, Orange lead

014232-A7 014232-A1
AA14232-A7EA 448 USD
AA14232-A7 AA14232-A1
Lead(II,IV) oxide ≥97% (metals basis)
Lead(II,IV) oxide

MDL: MFCD00078491


Purity: 97% CAS No.: 1314-41-6 Molecular Formula: O4Pb3 Formula Weight: 685.60 Physical Form: Powder Density: 9.1 Appearance: Bright red to orange Odor: Odorless Melting Point: ca 500° dec. Merck Reference: 14,5425 UN No.: 1479 MDL No.: MFCD00078491

Formula: Pb₃O₄
MW: 685.6 g/mol
Boiling Pt: 2682 °C (1013 hPa)
Melting Pt: 830 °C
Density: 9.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078491
CAS Number: 1314-41-6
EINECS: 215-235-6
UN: 1479
ADR: 5.1,II
Merck Index: 12,05449

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR