Tantalum pentachloride ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

014164-14 014164-22 014164-06
AA14164-14EA 162 USD
AA14164-14 AA14164-22 AA14164-06
Tantalum pentachloride ≥99.8% (metals basis)
Tantalum pentachloride

In chlorination of organic substances, as an intermediate, and in the production of pure metal


Purity: 99.8% CAS No.: 7721-01-9 Molecular Formula: TaCl5 Formula Weight: 358.21 Physical Form: Powder, Ampouled under argon Density: 3.68 Appearance: White to pale yellow Odor: Acidic Melting Point: 216° Boiling Point: 242° Solubility: Decomposes in water. Soluble in absolute alcohol and KOH Merck Reference: 14,9054 UN No.: 3260 MDL No.: MFCD00011253


In chlorination of organic substances, as an intermediate, and in the production of pure metal

Formula: Cl₅Ta
Boiling Pt: 242 °C
Melting Pt: 216 °C
Density: 3.68
Storage Temperature: Ambient
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III
Merck Index: 14,9054

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR