Lead titanate ≥99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead titanium oxide, Lead(II) titanium oxide, Lead(II) titanate, Lead titanate (IV), Lead titanate

014139-A3 014139-22 014139-36
AA14139-A3EA 250 USD
AA14139-A3 AA14139-22 AA14139-36
Lead titanate ≥99.5%
Lead titanate

MDL: MFCD00011159


Purity: 99.5% CAS No.: 12060-00-3 Molecular Formula: PbTiO3 Formula Weight: 303.09 Physical Form: -325 Mesh Powder Density: 7.520 Appearance: Yellow Odor: Odorless UN No.: 3288 MDL No.: MFCD00011159

Formula: O₃PbTi
MW: 303.08
Melting Pt: 1120
Density: 7.520
Storage Temperature: Ambient
MDL Number: MFCD00011159
CAS Number: 12060-00-3
EINECS: 235-038-9
UN: 3288
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR