Trichlorosilane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
014078-22
014078-36
AA14078-22EA
81.2
USD
AA14078-22
AA14078-36
Trichlorosilane 98%
Trichlorosilane
MDL: MFCD00011518
Soluble in benzene, CS2, chloroform, CCl4
Purity: 98%
CAS No.: 10025-78-2
Molecular Formula: HSiCl3
Formula Weight: 135.45
Storage Temperature: 2°C to 8°C
Flash Point: -13°(7°F)
Physical Form: Liquid
Density: 1.342
Refractive Index: 1.4020
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless
Melting Point: -127°
Boiling Point: 31-32°
Solubility: Soluble in benzene, CS2, chloroform, CCl4
Merck Reference: 14,9646
UN No.: 1295
MDL No.: MFCD00011518
Formula:
HCl₃Si MW: 135.45 g/mol Boiling Pt: 31…32 °C Melting Pt: –127 °C Density: 1.344 g/cm³ (20 °C) Flash Pt: –28 °C (7 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011518 CAS Number: 10025-78-2 EINECS: 233-042-5 UN: 1295 ADR: 4.3,I REACH: 01-2119494046-35 Merck Index: 12,09776 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...