Thulium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
013652-14
013652-06
AA13652-03EA
67.08
USD
AA13652-03
AA13652-14
AA13652-06
Thulium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Thulium(III) fluoride
MDL: MFCD00049611
Insoluble in water
Purity: 99.9%
CAS No.: 13760-79-7
Molecular Formula: TmF3
Formula Weight: 225.93
Physical Form: Crystalline
Density: 7.9
Melting Point: 1158°
Boiling Point: 2200°
Solubility: Insoluble in water
UN No.: 3288
MDL No.: MFCD00049611
Formula:
F₃Tm MW: 225.93 Boiling Pt: 2200 Melting Pt: 1158 Density: 7.9 Storage Temperature: Ambient |
MDL Number:
MFCD00049611 CAS Number: 13760-79-7 EINECS: 237-353-7 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...