Mercury (II) sulfide, red

Supplier: Thermo Scientific Chemicals
Warning

013482-22 013482-36
AA13482-22EA 221 USD
AA13482-22 AA13482-36
Mercury (II) sulfide, red
Mercury (II) sulphide

MDL: MFCD00011046


CAS No.: 1344-48-5 Molecular Formula: HgS Formula Weight: 232.65 Physical Form: Powder Density: 8.17 Appearance: Red Odor: Odorless Melting Point: 583.5° subl. Merck Reference: 14,5888 MDL No.: MFCD00011046

Formula: HgS
Melting Pt: 583.5 °C (subl.)
Density: 8.17
Storage Temperature: Ambient
MDL Number: MFCD00011046
CAS Number: 1344-48-5
EINECS: 215-696-3
Merck Index: 14,5888

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR