Digallium trisulfide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gallium (III) sulphide
013387-04
013387-02
013387-09
AA13387-04EA
110
USD
AA13387-04
AA13387-02
AA13387-09
Digallium trisulfide ≥99.99% (metals basis)
Digallium trisulphide
MDL: MFCD00016106
Purity: 99.99%
CAS No.: 12024-22-5
Molecular Formula: Ga2S3, may contain small amount of oxide
Formula Weight: 235.63
Physical Form: Powder
Density: 3.77
Melting Point: 1255°
UN No.: 3260
MDL No.: MFCD00016106
Formula:
Ga₂S₃ MW: 235.64 g/mol Melting Pt: 1255 °C Density: 3.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016106 CAS Number: 12024-22-5 EINECS: 234-692-2 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...