Copper(I) telluride ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
013140-06
013140-14
AA13140-06EA
75.05
USD
AA13140-06
AA13140-14
Copper(I) telluride ≥99.5% (metals basis)
Copper(I) telluride
MDL: MFCD00049727
Purity: 99.5%
CAS No.: 12019-52-2
Molecular Formula: Cu2Te
Formula Weight: 254.68
Physical Form: 3mm & down Pieces
Density: 7.27
Refractive Index: 1.45
Appearance: Blue-black
Odor: Odorless
MDL No.: MFCD00049727
Formula:
Cu₂Te MW: 254.69 Melting Pt: 1125 Density: 7.27 Storage Temperature: Ambient |
MDL Number:
MFCD00049727 CAS Number: 12019-52-2 EINECS: 234-646-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...