Sodium fluoride ≥99.995% (metals basis excluding K), Puratronic® (K ∼ 100 ppm)
Supplier: Thermo Scientific Chemicals
|
Danger
|
012964-30
012964-06
012964-18
AA12964-30EA
520
USD
AA12964-30
AA12964-06
AA12964-18
Sodium fluoride ≥99.995% (metals basis excluding K), Puratronic® (K ∼ 100 ppm)
Sodium fluoride
MDL: MFCD00003524
Fieser: 1,1073 2,382 6,473
Purity: 99.995%
CAS No.: 7681-49-4
Molecular Formula: NaF
Formula Weight: 41.99
Physical Form: Powder
Density: 2.78
Refractive Index: 1.336
Odor: Odorless
Melting Point: 993°
Boiling Point: 1704°
Merck Reference: 14,8618
UN No.: 1690
MDL No.: MFCD00003524
Formula:
NaF MW: 41.99 g/mol Boiling Pt: 1704 °C Melting Pt: 993 °C Density: 2.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 EINECS: 231-667-8 UN: 1690 ADR: 6.1,III Merck Index: 13,08691 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...