Rubidium nitrate ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
012889-18
012889-30
012889-09
AA12889-18EA
216
USD
AA12889-18
AA12889-30
AA12889-09
Rubidium nitrate ≥99.8% (metals basis)
Rubidium nitrate
MDL: MFCD00011193
Purity: 99.8%
CAS No.: 13126-12-0
Molecular Formula: RbNO3
Formula Weight: 147.47
Physical Form: Crystalline
Density: 3.11
Refractive Index: 1.52
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 310°
UN No.: 1477
MDL No.: MFCD00011193
Formula:
NO₃Rb MW: 147.47 Boiling Pt: 578 Melting Pt: 310 °C Density: 3.11 Storage Temperature: Ambient |
MDL Number:
MFCD00011193 CAS Number: 13126-12-0 EINECS: 236-060-1 UN: 1477 ADR: 5.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...