Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals
Warning

012426-FI 012426-FF
AA12426-FIEA 1872 USD
AA12426-FI AA12426-FF
Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.025 mm (0.001 in)
Scandium

MDL: MFCD00016323


Purity: 99.9% CAS No.: 7440-20-2 Physical Form: ≈0.05g/25x25mm, Packaged under argon Appearance: Grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00016323

Formula: Sc
MW: 44.96 g/mol
Boiling Pt: 2730 °C (1013 hPa)
Melting Pt: 1541 °C
Density: 2.989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
UN: 3178
ADR: 4.1,III
Merck Index: 12,08537

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR