Europium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Supplier: Thermo Scientific Chemicals
|
Danger
|
012394-06
012394-03
AA12394-06EA
756
USD
AA12394-06
AA12394-03
Europium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Europium
MDL: MFCD00010992
Notes: Due to the reactive nature of the material, it may contain darkened surface oxidation
Purity: 99.9%
CAS No.: 7440-53-1
Physical Form: Packaged under mineral oil
Sensitivity: Air and Moisture Sensitive
Appearance: Silver/gray
Odor: Odorless
UN No.: 3208
MDL No.: MFCD00010992
Formula:
Eu MW: 151.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010992 CAS Number: 7440-53-1 EINECS: 231-161-7 UN: 3208 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...