Mercury (II) oxide, yellow, Reagent Grade
Supplier: Thermo Scientific Chemicals
MDL: MFCD00011045
Fieser: 1,655 6,360 7,224 8,316 9,293 12,305 14,204 18,233 20,239
CAS No.: 21908-53-2
Molecular Formula: HgO
Formula Weight: 216.59
Physical Form: Powder
Density: 11.14
Odor: Odorless
Merck Reference: 14,5883
UN No.: 1641
MDL No.: MFCD00011045
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...