Ammonium dimolybdate Mo 56.5%

Supplier: Thermo Scientific Chemicals

012215-30 012215-A1 012215-18
AA12215-30EA 66.3 USD
AA12215-30 AA12215-A1 AA12215-18
Ammonium dimolybdate Mo 56.5%
Ammonium dimolybdate

MDL: MFCD00010882


CAS No.: 27546-07-2 Molecular Formula: (NH4)2Mo2O7 Formula Weight: 339.95 Physical Form: Powder MDL No.: MFCD00010882

Formula: H₈Mo₂N₂O₇
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010882
CAS Number: 27546-07-2
EINECS: 248-517-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR