Gold trichloride Au ≥ 64.4%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gold(III) chloride, Gold (III) chloride
012163-06
012163-03
012163-01
AA12163-06EA
913
USD
AA12163-06
AA12163-03
AA12163-01
Gold trichloride Au ≥ 64.4%
Gold trichloride
MDL: MFCD00014172
CAS No.: 13453-07-1
Molecular Formula: AuCl3
Formula Weight: 303.33
Physical Form: Crystalline
Density: 3.90
Sensitivity: Hygroscopic
Appearance: Red
Odor: Odorless
Melting Point: 180(subl)
Merck Reference: 14,4521
UN No.: 3260
MDL No.: MFCD00014172
Formula:
AuCl₃ MW: 303.32 g/mol Melting Pt: 180 °C (subl.) Density: 4.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014172 CAS Number: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8,III Merck Index: 13,04536 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...