Gold(III) oxide ≥99.99% (metals basis) Au ≥ 88.6%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

012161-03 012161-01 012161-06
AA12161-03EA 301 USD
AA12161-03 AA12161-01 AA12161-06
Gold(III) oxide ≥99.99% (metals basis) Au ≥ 88.6%, Premion®
Gold(III) oxide

MDL: MFCD00014173 Insoluble in water. Soluble in HCl2 HNO3, and in NaCN solution


Purity: 99.99% CAS No.: 1303-58-8 Molecular Formula: Au2O3 Formula Weight: 441.93 Physical Form: Powder Appearance: Brown Odor: Odorless Melting Point: 150° dec. Solubility: Insoluble in water. Soluble in HCl, HNO3, and in NaCN solution Merck Reference: 14,4524 MDL No.: MFCD00014173

Formula: Au₂O₃
MW: 441.93
Melting Pt: 150 (dec.)
Density: 3.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014173
CAS Number: 1303-58-8
EINECS: 215-122-1
Merck Index: 14,4524

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR