Vanadium carbide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

012141-22 012141-14
AA12141-22EA 185 USD
AA12141-22 AA12141-14
Vanadium carbide ≥99% (metals basis)
Vanadium carbide

MDL: MFCD00049698


Purity: 99% CAS No.: 12070-10-9 Molecular Formula: VC (VC0.88 by X-Ray) Formula Weight: 62.95 Physical Form: -325 Mesh Powder Odor: Odorless UN No.: 3178 MDL No.: MFCD00049698

Formula: CV
MW: 62.95
Boiling Pt: 3900
Melting Pt: 2810
Density: 5.77 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049698
CAS Number: 12070-10-9
EINECS: 235-122-5
UN: 3178
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR