Silver ≥99.9985% (metals basis), foil, Premion®, Thickness 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

012128-FF 012128-FI 012128-GH
AA12128-FFEA 63.4 USD
AA12128-FF AA12128-FI AA12128-GH
Silver ≥99.9985% (metals basis), foil, Premion®, Thickness 0.5 mm (0.02 in)
Silver

MDL: MFCD00003397


Purity: 99.9985% CAS No.: 7440-22-4 Physical Form: ≈3.3g/25x25mm Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR