Indium(III) iodide, anhydrous ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

011854-09 011854-18 011854-04
AA11854-09EA 282 USD
AA11854-09 AA11854-18 AA11854-04
Indium(III) iodide, anhydrous ≥99.999% (metals basis)
Indium(III) iodide

MDL: MFCD00016151


Purity: 99.999% CAS No.: 13510-35-5 Molecular Formula: I3In Formula Weight: 495.53 Physical Form: Crystalline Density: 4.69 Appearance: Yellow to brown Melting Point: 210° UN No.: 3260 MDL No.: MFCD00016151

Formula: I₃In
MW: 495.53 g/mol
Boiling Pt: 500
Melting Pt: 209.85
Density: 4.69
Storage Temperature: Ambient
MDL Number: MFCD00016151
CAS Number: 13510-35-5
EINECS: 236-839-6
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR