Barium perchlorate, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

011657-36 011657-22
AA11657-36EA 134 USD
AA11657-36 AA11657-22
Barium perchlorate, anhydrous
Barium perchlorate

Desiccant, ribonuclease determination


CAS No.: 13465-95-7 Molecular Formula: Ba(ClO4)2 Formula Weight: 336.24 Physical Form: Powder Density: 3.20 Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,987 UN No.: 1447 MDL No.: MFCD00003440


Desiccant, ribonuclease determination

Formula: Ba(ClO₄)₂
MW: 336.23 g/mol
Melting Pt: 505 °C
Density: 3.2 g/cm³ (20 °C)
Flash Pt: 21 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003440
CAS Number: 13465-95-7
EINECS: 236-710-4
UN: 1447
ADR: 5.1,II
Merck Index: 12,01016

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR