Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
011451-FY
011451-FG
AA11451-FYEA
742
USD
AA11451-FY
AA11451-FG
Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness 0.5 mm (0.02 in)
Cobalt
MDL: MFCD00010935
Purity: 99.995%
CAS No.: 7440-48-4
Physical Form: ≈5.6g/25x50mm
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...