Silver(I) oxide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

011407-22 011407-36 011407-14
AA11407-22EA 476 USD
AA11407-22 AA11407-36 AA11407-14
Silver(I) oxide ≥99% (metals basis)
Silver(I) oxide

Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent


Purity: 99+% CAS No.: 20667-12-3 Molecular Formula: Ag2O Formula Weight: 231.74 Physical Form: Powder Density: 7.2 Appearance: Dark brown Odor: Odorless Melting Point: 230° dec. Merck Reference: 14,8521 UN No.: 1479 MDL No.: MFCD00003404


Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent

Formula: Ag₂O
MW: 231.74 g/mol
Melting Pt: 230 °C (dec.)
Density: 7.2
Storage Temperature: Ambient
MDL Number: MFCD00003404
CAS Number: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1 II
Merck Index: 14,8521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR